UHPLC Analysis of Nifedipine on Titan™ C18
UHPLC Analysis of Nifedipine on Titan™ C18
Conditions
column Titan C18, 10 cm x 2.1 mm I.D., 1.9 µm particles (577124-U)
column temp. 35 °C
mobile phase [A] 10 mM ammonium acetate, pH 7.0; B] acetonitrile (70:30, A;B)
flow rate 0.3 mL/min
sample 300 ng/mL each in acetonitrile:water (3:1)
injection 2 µL
detector MS, XIC m/z 347.18
Description
Categories Analytical Chromatography, Antihypertensives, Calcium channel blockers
Featured Industry Pharmaceutical (small molecule)
Legal Information Titan is a trademark of Sigma-Aldrich Co. LLC
suitability application for UHPLC