UHPLC Analysis of Quinidine and Metabolite on Titan™ C18
UHPLC Analysis of Quinidine and Metabolite on Titan™ C18
Conditions
column Titan C18, 5 cm x 2.1 mm I.D., 1.9 µm particles (577122-U)
column temp. 35 °C
mobile phase [A] 10 mM potassium phosphate, monobasic pH 3.0; [B] acetonitrile; (80:20, A:B)
flow rate 0.6 mL/min
pressure 4310 psi (297 bar)
sample Quinidine 5 µg/mL in 80:20, water:methanol
injection 1 µL
detector UV, 200 nm
Description
Categories Analytical Chromatography, Antiarrhythmic drugs
Featured Industry Pharmaceutical (small molecule)
Legal Information Titan is a trademark of Sigma-Aldrich Co. LLC
suitability application for UHPLC