UHPLC Analysis of Stilbenes on Ascentis® Express C8 and ES-Cyano phases
UHPLC Analysis of Stilbenes on Ascentis® Express C8 and ES-Cyano phases
Conditions
column Ascentis Express ES-Cyano, 5 cm x 4.6 mm I.D., 5 µm particles (50581-U)
column Ascentis Express C8, 5 cm x 4.6 mm I.D., 5 µm particles (50389-U)
column temp. 30 °C
mobile phase [A] water; [B] acetonitrile
gradient 40-60% B in 3 minutes; hold at 60% B until 4 minutes
flow rate 2.0 mL/min
pressure 1740 psi (120 bar)
sample Dissolved in 50/50 water/ACN
injection 1 µL
detector UV, 230 nm
Description
Categories Analytical Chromatography, Antioxidants