Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
UHPLC Analysis of Vitamin K1 and Related Compound on Ascentis® Express C18
UHPLC Analysis of Vitamin K1 and Related Compound on Ascentis® Express C18
Conditions
column Ascentis Express C18 15 cm x 4.6 mm I.D., 5 µm particles (50537-U)
column temp. 35 C
mobile phase [A] methanol; [B] 2-propanol; (90:10, A:B)
flow rate 1.0 ml/min
pressure 1035 psi (71 bar)
sample Vitamin K1 and related, 50 µg/mL 90:10, methanol:2-propanol
injection 2 µl
detector UV, 250 nm
Description
Categories Analytical Chromatography, Vitamins and Natural products
Featured Industry Life Science and Biopharma
Legal Information Ascentis is a registered trademark of Sigma-Aldrich Co. LLC
suitability application for UHPLC