Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
UHPLC Gradient Analysis of Boswellic Acids on Titan™ C18
UHPLC Gradient Analysis of Boswellic Acids on Titan™ C18
Conditions
column Titan C18, 10 cm x 2.1 mm I.D., 1.9 µm particles (577124-U)
column temp. 35 C
mobile phase [A] 0.1% phosphoric acid; [B] acetonitrile
gradient 40% B for 0.5 min; to 100% B in 2 min; held at 100% B for 10 min
flow rate 0.4 ml/min
pressure 4050 psi (279 bar)
sample 6 Boswellic Acids 50µg/mL in 70:30, water:methanol
injection 2 µl
detector UV, 210 nm
Description
Categories Analytical Chromatography, Herbal medicines
Featured Industry Vitamins, Nutraceuticals, and Natural Products
Legal Information Titan is a trademark of Sigma-Aldrich Co. LLC
suitability application for UHPLC