UHPLC Gradient Analysis of Boswellic Acids on Ascentis® Express Phenyl-Hexyl
UHPLC Gradient Analysis of Boswellic Acids on Ascentis® Express Phenyl-Hexyl
Conditions
column Ascentis Express Phenyl-Hexyl, 10 cm x 3.0 mm I.D., 2.7 µm particles (53345-U)
column temp. 35 C
mobile phase [A] 0.1% phosphoric acid; [B] acetonitrile
gradient 50% B for 0.5 min; to 100% B in 4.5 min; held at 100% B for 5 min
flow rate 0.6 ml/min
pressure 2390 psi (165 bar)
sample 6 Boswellic Acids 50µg/mL in 70:30, water:methanol
injection 2 µl
detector UV, 210 nm
Description
Categories Analytical Chromatography, Herbal medicines
Featured Industry Vitamins, Nutraceuticals, and Natural Products
Legal Information Ascentis is a registered trademark of Sigma-Aldrich Co. LLC
suitability application for UHPLC