UHPLC/MS Analysis of Cannabinoids Extracted from Raw Cannabis Sativa (Marijuana) on Ascentis® Express RP-Amide
UHPLC/MS Analysis of Cannabinoids Extracted from Raw Cannabis Sativa (Marijuana) on Ascentis® Express RP-Amide
Conditions
sample preparation A 100 mg sample of dried cannabis sativa leaves was extracted with 50 mL of ethanol:water (50:50, v/v) for 15 mins in an ultrasonic bath. (The sample was then filtered through a 0.45 µm Millex LH filter prior to LC/MS analysis.)
column Ascentis Express RP-Amide, 10 cm x 2.1 mm I.D., 2.0 µm particles (53913-U)
column temp. 35 C
mobile phase [A] 0.1% formic acid; [B] 0.1% formic acid in acetonitrile
gradient 60% B to 100% B in 10 min
flow rate 0.4 ml/min
pressure 7600 psi (524 bar)
injection 1 µl
detector MS, ESI(+), ESI(-), MRM
Description
Analysis Note Cannabis compounds reportedly have therapeutic efficacy in the treatment of pain, mood disorders, inflammatory diseases, and other health conditions. Shown here is a raw cannabis sample extracted using a simple procedure followed by LC/MS/MS analysis. Eight major cannabis compounds were detected. Reversed-phase HPLC separation was achieved on an Ascentis Express RP-Amide column. Dried cannabis sample was obtained courtesy of Dr. Hari H. Singh, Program Director at the Chemistry & Physiological Systems Research Branch of the National Institute on Drug Abuse at the US National Institute of Health. The extract strain of the sample was not known.
Categories Analytical Chromatography, Pharmaceuticals, Herbal medicines, Drugs of Abuse
Featured Industry Food and Beverages
Forensics and Toxicology
Vitamins, Nutraceuticals, and Natural Products
Legal Information Ascentis is a registered trademark of Sigma-Aldrich Co. LLC
suitability application for UHPLC-MS