UHPLC/MS/MS Analysis of 25 Small Molecule Pharmaceutical Compounds on Ascentis® Express RP-Amide
UHPLC/MS/MS Analysis of 25 Small Molecule Pharmaceutical Compounds on Ascentis® Express RP-Amide
Conditions
column Ascentis Express RP-Amide, 10 cm x 2.1 mm I.D., 2.7 µm particles (53913-U)
mobile phase [A] 0.1% acetic acid, 5 mM ammonium acetate in water; [B] 5 mM ammonium acetate in acetonitrile
gradient 5% B for 1 min; to 43% B in 6 min; held for 1 min; to 80% B in 4 min; held for 1 min; to 5% B in 1 min; held for 3 min
flow rate 0.4 ml/min
injection 1 µl
detector MS, ESI(+) MRM
Description
Antigen Background This application highlights the advantage of an embedded polar group (EPG) HPLC phase for the separation of various small molecules. The use of an Ascentis® RP-Amide column allowed for the development of a single UPLC-MS/MS method for the analysis of 25 pharmaceuticals that varied significantly in both chemical and physical properties (log P, pKa, etc.). The presence of an embedded amide group allowed for retention of more polar analytes in comparison to a traditional C18 phase.
Categories Analytical Chromatography, Pharmaceuticals
Featured Industry Pharmaceutical (small molecule)
Legal Information Ascentis is a registered trademark of Sigma-Aldrich Co. LLC
suitability application for UHPLC-MS