Higher Efficiency of Ascentis® Express 2.7 μm Compared to 3 μm Particles Gives Better Sensitivity
Higher Efficiency of Ascentis® Express 2.7 μm Compared to 3 μm Particles Gives Better Sensitivity
Conditions
column Ascentis Express C18, 5 cm x 2.1 mm I.D., 2.7 µm particles and C18, 5 cm x 2.1 mm I.D., 3 µm particles (53822-U)
column temp. 35 °C
mobile phase [A] 25 mM dibasic ammonium phosphate (pH 7.0): [B] water: [C] acetonitrile; (35:0:65 or 35:4:61, A:B:C)
flow rate 0.2 mL/min
injection 1 µL
detector UV, 220 nm
Description
Categories Analytical Chromatography, Pharmaceutical Analysis, Antiarrythmic, Antidepressants, Antihistamines