US EPA Method 632: HPLC Analysis of Pesticides on Ascentis® C18
US EPA Method 632: HPLC Analysis of Pesticides on Ascentis® C18
Conditions
column Ascentis C18, 25 cm x 4.6 mm I.D., 5 µm particles (581325-U)
column temp. 30 C
mobile phase [A] water; [B] acetonitrile
gradient 10 to 50% B in 13 min; to 60% B in 10 min; to 90% B in 6 min; held for 5 min
flow rate 1.0 ml/min
sample as indicated in 16% acetonitrile in water
injection 10 µl
detector UV, 210 nm
Description
Categories Analytical Chromatography, Environmental Analysis, Organics, Pesticides & Herbicides, Chromatograms, HPLC Chromatograms
Featured Industry Agriculture
Environmental