US EPA Method 632: HPLC Analysis of Pesticides on Ascentis® C8
US EPA Method 632: HPLC Analysis of Pesticides on Ascentis® C8
Conditions
column Ascentis C8, 25 cm x 4.6 mm I.D., 5 µm particles (581425-U)
column temp. 30 °C
mobile phase [A] DI water; [B] acetonitrile
gradient 15 to 25% B in 4 min; to 45% B in 40 min; to 60% B in 6 min; held for 15 min
flow rate 1.0 mL/min
sample as indicated in 16% acetonitrile in DI water
injection 10 µL
detector UV, 210 nm
Description
Categories Analytical Chromatography, Environmental Analysis, Organics, Pesticides & Herbicides, Chromatograms, HPLC Chromatograms
Featured Industry Environmental