US EPA Method 8315 (Solid Waste/Ground Water Carbonyls by HPLC-UV) on Ascentis® C18
US EPA Method 8315 (Solid Waste/Ground Water Carbonyls by HPLC-UV) on Ascentis® C18
Conditions
column Ascentis C18, 15 cm x 4.6 mm I.D., 3 µm particles (581322-U)
column temp. 30 °C
mobile phase 30:70 water:acetonitrile
flow rate 1 mL/min
sample 15-component DNPH-derivatized carbonyl mix, each analyte at15 µg/mL (aldehyde equivalent) in acetonitrile (47285-U)
injection 1 µL
detector UV, 365 nm
Description
Analysis Note This application demonstrates the suitability of the Ascentis C18 for the analysis of 15 aldehyde/ketone DNPH derivatives.
Featured Industry Environmental