Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
US EPA Method 8330: HPLC Analysis of Nitroaromatics and Nitramines, Mix A on Ascentis® C18
US EPA Method 8330: HPLC Analysis of Nitroaromatics and Nitramines, Mix A on Ascentis® C18
Conditions
column Ascentis C18, 25 cm x 2.1 mm I.D., 5 µm particles (581305-U)
column temp. 60 C
mobile phase (A) water: (B) methanol; (50:50, A:B)
flow rate 0.2 ml/min
sample acetonitrile
injection 1 µl
detector UV, 254 nm
Description
Categories Analytical Chromatography, Environmental Analysis, Explosives, Air toxics, Organics, Chromatograms, HPLC Chromatograms
Featured Industry Environmental