US EPA Method 8330: HPLC Analysis of Nitroaromatics and Nitroamines, Mix B on Ascentis® C18
US EPA Method 8330: HPLC Analysis of Nitroaromatics and Nitroamines, Mix B on Ascentis® C18
Conditions
column Ascentis C18, 25 cm x 2.1 mm I.D., 5 µm particles (581305-U)
column temp. 60 °C
mobile phase (A) water;(B) methanol;(50:50, A:B)
flow rate 0.2 mL/min
sample as indicated in acetonitrile
injection 1 µL
detector UV, 254 nm
Description
Categories Analytical Chromatography, Environmental Analysis, Explosives, Air toxics, Organics, Chromatograms, HPLC Chromatograms
Featured Industry Environmental