USP Analysis of Actos (Pioglitazone) using Ascentis® Express C18
USP Analysis of Actos (Pioglitazone) using Ascentis® Express C18
Conditions
column Ascentis® Express C18, 15 cm x 4.6 mm, 5 µm particles (50537-U)
column temp. 25 °C
mobile phase [A] 100 mM ammonium acetate; [B] acetonitrile; [C] acetic acid; (55:45:2, A:B:C)
flow rate 0.7 mL/min
pressure 1160 psi (80 bar)
sample pioglitazone 50 µg/mL, benzophenone 13 µg/mL in mobile phase
injection 20 µL
detector UV, 269 nm
Description
Categories Analytical Chromatography, Antihyperglycemic