USP HPLC Analysis of Ampicillin Sodium on Ascentis® Express C18
USP HPLC Analysis of Ampicillin Sodium on Ascentis® Express C18
Conditions
column Ascentis Express C18, 10 cm x 4.6 mm I.D., 5 mum particles (50536-U)
column temp. 30 °C
mobile phase [A:B:C:D] [A] acetonitrile, [B]water, [C] 1M Potassium phosphate monobasic in water, [D] 1N acetic acid in water (80:909:10:1)
flow rate 1.0 mL/min
sample 1 mg/mL in mobile phase
injection 10 µL
detector UV, 254 nm
Description
Analysis Note This application was generated by RTC on an Ascentis Express C18.
Categories Analytical Chromatography, Antibiotics, Antimicrobials