USP Method for the HPLC Analysis of 4-Amino-6-Chloro-1,3-Benzenedisulfonamide on Ascentis® Express C18
USP Method for the HPLC Analysis of 4-Amino-6-Chloro-1,3-Benzenedisulfonamide on Ascentis® Express C18
Conditions
column Ascentis Express C18, 5 cm x 4.6 mm I.D., 2.7 µm particles (53826-U)
column temp. 30 °C
mobile phase [A] formic acid in water (5:1000); [B] acetonitrile:methanol (3:1)
gradient 3% B for 5 min; to 36% B in 14 min; to 3% B in 18 min
flow rate 1.0 mL/min
pressure 1189 psi (82 bar)
sample 0.32 mg/mL in 2.76 mg/mL sodium phosphate, monobasic (pH 2.7 with phosphoric acid):acetonitrile (7:3)
injection 6 µL
detector UV, 275 nm
Description
Categories Analytical Chromatography, Diuretics