Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Determination of Water Content in Dry fruits Using Karl Fischer Titration
Product Group
Foodstuffs
General Information concerning the product group
Foodstuffs
Foodstuffs encompass the most diverse products. Depending on whether carbohydrate-rich, fatty or proteinrich substances are under investigation, different working techniques are favourable. Complex matrices that dissolve slowly, or where the water can only be slowly extracted, necessitate the use of solubilisers. In addition, titration under heating and the application of a homogeniser to accelerate water release are expedient.
Special Information concerning the sample and the methods
The dissolution of carbohydrates in methanolic KF solvents is often inadequate. For complete dissolution of the sample the addition of formamide is necessary. However, the proportion of formamide should not exceed 50 %. Additionally, titration under warm conditions (50 °C) is expedient. Dry fruits may be titrated both, at room temperature as well as at 50 °C.
Titration one component system
Reagents:
Titrant
188005 Aquastar™ - CombiTitrant 5 - One component reagent for volumetric Karl Fischer titration, 1 mL = approx. 5 mg water
Solvent
30 mL 188009 Aquastar™ - CombiMethanol - Solvent for volumetric Karl Fischer titration with one component reagents, max. 0.01 % water
and
20 mL 109684 Formamide - as Solubilizer
Titration Parameters:
Stirring time: 180 sec.
Default titration settings, e.g.:
I(pol) = 20 - 50 µA, U(EP) = 100 - 250 mV
Stop criterion: drift < 20 µL/min
Sample size:
0,1 g (powdered)
Procedure:
The titration medium is first placed into the titration cell and titrated dry by means of the titrant. Then the sample is added from a weighing boat (exact sample weight determination by weighing of weighing boat before and after addition) and the titration is started. For complete extraction of the sample a stirring time of at least 180 seconds is recommended.
Titration two component system
Reagents:
Titrant
188010 Aquastar™ - Titrant 5 - Titrant for volumetric titration with two component reagents, 1 mL = approx. 5 mg water
Solvent
30 mL 188015 Aquastar™ - Solvent - Solvent for volumetric titration with two component reagents
and
20 mL 109684 Formamide - as Solubilizer
Titration Parameters:
Stirring time: 180 sec.
Default titration settings, e.g.: z.B.
I(pol) = 20 - 50 µA, U(EP) = 100 - 250 mV
Stop criterion: drift < 20 µL/min
Sample size:
0,1 g (powdered)
Procedure:
The titration medium is first placed into the titration cell and titrated dry by means of the titrant. Then the sample is added from a weighing boat (exact sample weight determination by weighing of weighing boat before and after addition) and the titration is started. For complete extraction of the sample a stirring time of at least 180 seconds is recommended.