Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Determination of Water Content in Esomeprazole Using Karl Fischer Titration
Product Group
Pharmaceuticals
General Information concerning the product group
Pharmaceuticals
Pharmaceutical products are often characterized by complex formulations. Difficulties observed during Karl Fischer determination are often caused by the limited solubility. In some cases side reactions have to be considered. In dependance of composition and properties of the formulations, various measures are necessary for an undisturbed Karl Fischer determination. In pharmaceutical guidelines (USP, Ph Eur, DAB) the Karl Fischer titration is described as common method for water determination. For some substances special procedures can be found. The determination of mass loss as method for water determination is not recommended.
Special Information concerning the sample and the methods
Water determination in Esomeprazole can be carried out without problems according to standard methods.
Titration one component system
Reagents:
Titrant
188005 Aquastar™ - CombiTitrant 5 - One component reagent for volumetric Karl Fischer titration, 1 mL = approx. 5 mg water
Solvent
50 ml 188008 Aquastar™ - CombiSolvent - Methanol-free solvent for volumetric Karl Fischer titration with one component reagents
Titration Parameters:
Stirring time: 90 s
Default titration settings, e.g.:
I(pol) = 20 - 50 µA, U(EP) = 100 - 250 mV
Stop criterion: drift < 20 µL/min
Sample size:
0.2 g
Procedure:
The titration medium is first placed into the titration cell and titrated dry by means of the titrant. Then the sample is added from a weighing boat (exact sample weight determination by weighing of weighing boat before and after addition) and the titration is started. For complete dissolution of the sample a stirring time of 90 seconds is recommended.