Determination of Water Content in Polysorbate 80 (Tween 80) Using Karl Fischer Titration
Product Group
Cosmetics, Foodstuffs, Pharmaceuticals
General Information concerning the product group
Cosmetics
Cosmetics on an aqueous and alcoholic basis can be titrated without problems according to the Karl Fischer (KF) method. Due to the often high water concentration of the emulsions small samples are required. The accuracy of analysis is determined through exact sampling, weighing and adding. The coulometric method is unsuitable for most emulsions. Fatty and oily preparations are insoluble in methanolic KF solvents and require the addition of a solubiliser, or respectively titration with specific KF solvents for oils and fats.
Foodstuffs
Foodstuffs encompass the most diverse products. Depending on whether carbohydrate-rich, fatty or proteinrich substances are under investigation, different working techniques are favourable. Complex matrices that dissolve slowly, or where the water can only be slowly extracted, necessitate the use of solubilisers. In addition, titration under heating and the application of a homogeniser to accelerate water release are expedient.
Pharmaceuticals
Pharmaceutical products are often characterized by complex formulations. Difficulties observed during Karl Fischer determination are often caused by the limited solubility. In some cases side reactions have to be considered. In dependance of composition and properties of the formulations, various measures are necessary for an undisturbed Karl Fischer determination. In pharmaceutical guidelines (USP, Ph Eur, DAB) the Karl Fischer titration is described as common method for water determination. For some substances special procedures can be found. The determination of mass loss as method for water determination is not recommended.
Special Information concerning the sample and the methods
Water determination in Polysorbate 80 (Tween 80) can be carried out according to standard methods.
Titration one component system
Reagents:
Titrant
188005 Aquastar™ - CombiTitrant 5 - One component reagent for volumetric Karl Fischer titration, 1 mL = approx. 5 mg water
Solvent
50 mL 188009 Aquastar™ - CombiMethanol - Solvent for volumetric Karl Fischer titration with one component reagents, max. 0.01 % water
Titration Parameters:
Default titration settings, e.g.:
I(pol) = 20 - 50 µA, U(EP) = 100 - 250 mV
Stop criterion: drift < 20 µL/min
Sample size:
1 g
Procedure:
The titration medium is first placed into the cell and titrated dry by means of the titrant. Then the sample is added with a syringe (exact sample weight determination by weighing of syringe before and after injection) and the titration is started.
Titration two component system
Reagents:
Titrant
188010 Aquastar™ - Titrant 5 - Titrant for volumetric titration with two component reagents, 1 mL = approx. 5 mg water
Solvent
50 mL 188015 Aquastar™ - Solvent - Solvent for volumetric titration with two component reagents
Titration Parameters:
Default titration settings, e.g.: z.B.
I(pol) = 20 - 50 µA, U(EP) = 100 - 250 mV
Stop criterion: drift < 20 µL/min
Sample size:
1 g
Procedure:
The titration medium is first placed into the titration cell and titrated dry by means of the titrant. Then the sample is added with a syringe (exact sample weight determination by weighing of syringe before and after injection) and the titration is started.
Coulometry without diaphragm
Reagents:
Working medium
100 mL 109257 Aquastar™ - CombiCoulomat fritless - Coulometric Karl Fischer reagent for cells with or without diaphragm
Titration Parameters:
Default coulometer settings for cell without diaphragm:
For end point indication, e.g.:
I(pol) = 5 - 10 µA, U(EP) = 50 - 100 mV
Stop criterion for fast titration: drift < 20 µg/min
Sample size:
0,5 g
Procedure:
The Karl-Fischer reagent is placed into the titration cell without diaphragm. The coulometer is started and the solvent is titrated dry. After preliminary titration and stabilisation of drift the sample is injected into the titration cell with a syringe (exact sample weight determination by weighing of syringe before and after injection) and the water determination is started.