Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Analytical Method: Ammonium in mineral fertilizer
Instruments and Reagents:
Cat.No. 173016  Spectroquant® VIS Spectrophotometer Prove 100 or
Cat.No. 173017  Spectroquant® UV/VIS Spectrophotometer Prove 300 or
Cat.No. 173018  Spectroquant® UV/VIS Spectrophotometer Prove 600 or
Cat.No. 109748  Spectroquant® Photometer NOVA 30 or
Cat.No. 109751  Spectroquant® Photometer NOVA 60 or
Cat.No. 173632  Spectroquant® Colorimeter Move 100
Cat.No. 114946  Rectangular cells 10 mm and/or
Cat.No. 114947  Rectangular cells 20 mm and/or
Cat.No. 114944  Rectangular cells 50 mm
Cat.No. 114544  Spectroquant® Ammonium Cell Test*
Cat.No. 114559  Spectroquant® Ammonium Cell Test
Cat.No. 114752  Spectroquant® Ammonium Test
Cat.No. 100683  Spectroquant® Ammonium Test
Cat.No. 109057  Hydrochloric acid 1 mol/l
Cat.No. 116754  Water for analysis
Folded filter
*not compatible with Move 100
Sample preparation:
In a mill grind down about 30 g of the sample to a fine powder and homogenize. Bring to the boil 0.5 g of the powdered material with 25 ml water for analysis and 2.5 ml hydrochloric acid 1 mol/l. After cooling filter the mixture through a folded filter, wash the filter 3 times with boiling water for analysis and collect the filtrate and the washings in a 500-ml volumetric flask. Fill up to volume with water for analysis and mix. Finally put 10 ml of this solution into a 1000-ml volumetric flask, fill up to volume and mix.
Analysis:
Determine with the above mentioned test kits.
Calculation:
Ammonium content in % NH4 = analysis value in mg/l NH4 x 10
Materials
Related Links
Categories