Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Analytical Method: Analytical Quality Assurance, Standard for Calcium
Reagents:
Cat.No. 102121  Calcium nitrate tetrahydrate for analysis
Cat.No. 116754  Water for analysis
Preparation of a standard solution:
Dissolve 2.946 g of Calcium nitrate tetrahydrate in a calibrated or conformity-checked 500-ml volumetric flask and make up to the mark with distilled water. The stock solution prepared according to this procedure has a concentration of 1000 mg/l calcium.
Further investigational concentrations may be prepared from this stock solution by diluting accordingly.
Stability:
The stock solution of 1000 mg/l remains stable for one week. The diluted investigational solutions remain stable for one day.
For use with test kits:
Measuring range
in mg/l Ca
110083
MQuantTM Calcium Test
10 - 100
111110
MColortestTM Calcium Cell Test
2 - 200
116993
Reflectoquant® Calcium Test
2.5 - 45.0
116125
Reflectoquant® Calcium Test
5 - 125
100858
Spectroquant® Calcium Cell Test
10 - 250
100049
Spectroquant® Calcium Test
0.20 - 4.00
114815
Spectroquant® Calcium Test
5 - 160