Analytical Method: Analytical Quality Assurance, Standard for COD
Reagents:
Cat.No. 102400  Potassium hydrogen phthalate for analysis, volumetric standard
Cat.No. 116754  Water for analysis
Preparation of a standard solution:
Dissolve 0.851 g of potassium hydrogen phthalate in a calibrated or conformity-checked 1000-ml volumetric flask and make up to the mark with distilled water. The stock solution prepared according to this procedure has a concentration of 1000 mg/l COD. Further investigational concentrations may be prepared from this stock solution by diluting accordingly.
Stability:
When stored in a cool place (refrigerator), the stock solution remains stable for one month. When stored under appropriate cool conditions (refrigerator), the diluted investigational solutions remain stable - depending on the respective concentration - for approximately one week to one month.
For use with test kits:
Measuring range
in mg/l COD
114560
Spectroquant® COD Cell Test
4.0 - 40.0
101796
Spectroquant® COD Cell Test
5.0 - 80.0
114540
Spectroquant® COD Cell Test
10 - 150
114895
Spectroquant® COD Cell Test
15 - 300
114690
Spectroquant® COD Cell Test
50 - 500
114541
Spectroquant® COD Cell Test
25 - 1500
114691
Spectroquant® COD Cell Test
300 - 3500
114555
Spectroquant® COD Cell Test
500 - 10 000
109772
Spectroquant® COD Cell Test (Hg free)
10 - 150
109773
Spectroquant® COD Cell Test (Hg free)
100 - 1500
118750
Spectroquant® COD Cell Test for photometers of other manufacturers
0 - 40
118751
Spectroquant® COD Cell Test for photometers of other manufacturers
0 - 150
118752
Spectroquant® COD Cell Test for photometers of other manufacturers
0 - 1500
118753
Spectroquant® COD Cell Test for photometers of other manufacturers
0 - 15000