Analytical Method: Analytical Quality Assurance, Standard for Free Chlorine (dichloroisocyanuric acid)
Preparation of a standard solution for free chlorine
Reagents:
Cat.No. 110888  Dichloroisocyanuric acid sodium salt for analysis
Cat.No. 116754  Water for analysis
Preparation of a standard solution:
Dissolve 1.850 g of dichloroisocyanuric acid sodium salt with water for analysis in a calibrated or conformity-checked 1000-ml volumetric flask and make up to the mark with water for analysis. The stock solution prepared according to this procedure has a concentration of 1000 mg/l free chlorine.
Further investigational concentrations may be prepared from this stock solution by diluting accordingly.
Stability:
When stored in a cool place (refrigerator), the stock solution and the diluted investigational solutions remain stable for one day.
For use with test kits:
Measuring range
in mg/l Cl2
117925
MQuantTM Chlorine Test (free chlorine)
4 - 120
117924
MQuantTM Chlorine Test (free chlorine)
25 - 500
114434
MColortestTM Chlorine Test (free chlorine)
0.01 - 0.3
111174
MColortestTM Chlorine and pH Test (free + total chlorine)
0.1 - 1.5
111113
MColortestTM Chlorine Test Reagent Cl2-1
111114
MColortestTM Chlorine Test Reagent Cl2-1
111160
MColortestTM Chlorine and pH Test (free chlorine)
0.1 - 1.5
For use with test kits:
Measuring range
in mg/l
114670
MColortestTM Chlorine Test (free chlorine)
0.1 - 2
114978
MColortestTM Chlorine Test (free chlorine, liquid)
0.1 - 2
114979
MColortestTM Chlorine Test (free chlorine, liquid)
refill pack for 114978 
114801
MColortestTM Chlorine Test (free + total chlorine, liquid)
0.1 - 2
114803
MColortestTM Chlorine Test (free + total chlorine, liquid)
refill pack for 114801
114976
MColortestTM Chlorine Test (free chlorine)
0.25 - 15
114977
MColortestTM Chlorine Test (free chlorine)
refill pack for 114976
114826
MColortestTM Chlorine Test (free + total chlorine)
0.25 - 15
118326
MColortestTM Chlorine Test (free + total chlorine)
refill pack for 114826
117940
Reflectoquant® Chlorine Test (free + total chlorine)
0.05 - 2.0
116896
Reflectoquant® Chlorine Test (free + total chlorine)
0.5 - 10.0
100595
Spectroquant® Chlorine Cell Test (free chlorine)
0.03 - 6.00
100598
Spectroquant® Chlorine Test (free chlorine)
0.010 - 6.00
100086
Spectroquant® Chlorine Reagents Cl2-1
0.010 - 6.00
100087
Spectroquant® Chlorine Reagents Cl2-2
0.010 - 6.00
119254
Spectroquant® Chlorine Powder Packs for non-Merck photometers for 10-ml-samples (free chlorine)
0 - 2.00
119256
Spectroquant® Chlorine Powder Packs for non-Merck photometers for 25-ml-samples (free chlorine)
0 - 10.00
100597
Spectroquant® Chlorine Cell Test (free + total chlorine)
0.03 - 6.00
100599
Spectroquant® Chlorine Test (free + total chlorine)
0.010 - 6.00