Analytical Method: Analytical Quality Assurance, Standard for Free Chlorine (DIN)
Reagents:
Cat.No. 102404  Potassium iodate volumetric standard
Cat.No. 105043  Potassium iodide for analysis
Cat.No. 109072  Sulfuric acid 0.5 mol/l
Cat.No. 109136  Sodium hydroxide solution 2 mol/l
Cat.No. 116754  Water for analysis
Preparation of a KlO3 stock solution:
Dissolve 1.006 g of KlO3 in 250 ml of water for analysis in a calibrated or conformity-checked 1000-ml volumetric flask. Subsequently make up to the mark with water for analysis.
Preparation of a KlO3/Kl standard solution:
Transfer 15.0 ml of the KlO3 stock solution to a calibrated or conformity-checked 1000-ml volumetric flask, add approx. 1 g of Kl and make up to the mark with water for analysis. 1 ml of this solution is equivalent to 0.015 mg of free chlorine.
Preparation of the chlorine standard solution:
Pipette 20.0 ml KlO3/Kl standard solution into a calibrated or conformity-checked 100-ml volumetric flask, add
2 ml of H2SO4  0.5 mol/l, leave to stand for 1 min, and then add NaOH 2 mol/l dropwise (approx. 1 ml) until the solution just loses its color. Subsequently make up the solution to the mark with water for analysis. The concentration of the solution is 3.00 mg/l free chlorine.
Stability:
The KlO3 stock solution remains stable for 4 weeks when stored in a cool place. The KlO3/Kl standard solution can be used for 5 hours when stored in a cool place. The dilute iodine standard solution is not stable and must be used immediately.
For use with test kits:
Measuring range
in mg/l Cl2
117925
MQuantTM Chlorine Test (free chlorine)
4 - 120
117924
MQuantTM Chlorine Test (free chlorine)
25 - 500
114434
MColortestTM Chlorine Test (free chlorine)
0.01 - 0.3
111174
MColortestTM Chlorine and pH Test (free + total chlorine)
0.1 - 1.5
111157
MColortestTM Chlorine and pH Test (free + total chlorine) refill pack for 111174
111113
MColortestTM Chlorine Test Reagent Cl2-1
111114
MColortestTM Chlorine Test Reagent Cl2-2
111160
MColortestTM Chlorine and pH Test (free chlorine)
0.1 - 1.5
114670
MColortestTM Chlorine Test (free chlorine)
0.1 - 2
114978
MColortestTM Chlorine Test (free chlorine, liquid)
0.1 - 2
114979
MColortestTM Chlorine Test (free chlorine, liquid)
refill pack for 114978 
1.14801
MColortestTM Chlorine Test (free + total chlorine, liquid)
0.1 - 2
114803
MColortestTM Chlorine Test (free + total chlorine, liquid)
refill pack for 114801
114976
MColortestTM Chlorine Test (free chlorine)
0.25 - 15
114977
MColortestTM Chlorine Test (free chlorine)
refill pack for 114976
114826
MColortestTM Chlorine Test (free + total chlorine)
0.25 - 15
118326
MColortestTM Chlorine Test (free + total chlorine)
refill pack for 114826
116896
Reflectoquant® Chlorine Test (free + total chlorine)
0.5 - 10.0
100595
Spectroquant® Chlorine Cell Test (free chlorine)
0.03 - 6.00
100598
Spectroquant® Chlorine Test (free chlorine)
0.010 - 6.00
100086
Spectroquant® Chlorine Reagents Cl2-1
0.010 - 6.00
100087
Spectroquant® Chlorine Reagents Cl2-2
0.010 - 6.00
119254
Spectroquant® Chlorine Powder Packs for non-Merck photometers for 10-ml-samples (free chlorine)
0 - 2.00
119256
Spectroquant® Chlorine Powder Packs for non-Merck photometers for 25-ml-samples (free chlorine)
0 - 10.00
100597
Spectroquant® Chlorine Cell Test (free + total chlorine)
0.03 - 6.00
100599
Spectroquant® Chlorine Test (free + total chlorine)
0.010 - 6.00