Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Analytical Method: Analytical Quality Assurance, Standard for Hydrazine
Reagents:
Cat.No. 104603  Hydrazinium sulfate for analysis
Cat.No. 116754  Water for analysis
Preparation of a standard solution:
Dissolve 4.07 g of hydrazinium sulfate with oxygen-low (boil previously) distilled water in a calibrated or conformity-checked 1000-ml volumetric flask and make up to the mark with distilled water. The stock solution prepared according to this procedure has a concentration of 1000 mg/l hydrazine.
Further investigational concentrations may be prepared from this stock solution by diluting accordingly.
Stability:
When stored in a cool place (refrigerator), the stock solution and the further diluted investigational solutions remain stable for one day.
For use with test kits:
Measuring range
in mg/l
108017
MColortestTM Hydrazine Test
0.1 - 1 N2H4
109711
Spectroquant® Hydrazine Test
0.005 - 2.00 N2H4