Analytical Method: Analytical Quality Assurance, Standard for Magnesium
Reagents:
Cat.No. 105853  Magnesium nitrate hexahydrate for analysis
Cat.No. 116754  Water for analysis
Preparation of a standard solution:
Dissolve 1.055 g of magnesium nitrate hexahydrate in a calibrated or conformity-checked 100-ml volumetric flask and make up to the mark with distilled water. The stock solution prepared according to this procedure has a concentration of 1000 mg/l magnesium.
Further investigational concentrations may be prepared from this stock solution by diluting accordingly.
Stability:
The stock solution of 1000 mg/l remains stable for one week. The diluted investigational solutions remain stable for one day.
For use with test kits:
Measuring range
in mg/l Mg
111131
MColortestTM Magnesium Test
100 - 1500
116124
Reflectoquant® Magnesium Test
5 - 100
100815
Spectroquant® Magnesium Test
5.0 - 75.0