Analytical Method: Analytical Quality Assurance, Standard for Nitrogen (total)
Reagents:
Cat.No. 104201  Glycine for analysis
Cat.No. 116754  Water for analysis
Preparation of a standard solution:
Dissolve 5.36 g of glycine with distilled water in a calibrated or conformity-checked 1000-ml volumetric flask and make up to the mark with distilled water. The stock solution prepared according to this procedure has a concentration of 1000 mg/l total nitrogen. Further investigational concentrations may be prepared from this stock solution by diluting accordingly.
Stability:
When stored in a cool place (refrigerator), the stock solution remains stable for one week. The diluted investigational concentrations must be used immediately.
For use with test kits:
Measuring range
in mg/l
100613
Spectroquant® Nitrogen (total) Cell Test
0.5 - 15.0 N
114537
Spectroquant® Nitrogen (total) Cell Test
0.5 - 15.0 N
114763
Spectroquant® Nitrogen (total) Cell Test
10 - 150 N