Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Analytical Method: Analytical Quality Assurance, Standard for Sodium
Reagents:
Cat.No. 119897  Chloride standard solution 1000 mg/l Certipur®
Cat.No. 116754  Water for analysis
Preparation of a standard solution:
A chloride standard solution 1000 mg/l chloride can be used; 1000 mg/l chloride corresponds to 649 mg/l sodium.
Further investigational concentrations may be prepared from this stock solution by diluting accordingly.
Stability:
When stored in a cool place (refrigerator), the stock solution remains stable for one month.
For use with test kits:
Measuring range
in mg/l Na
100885
Spectroquant®Sodium Cell Test in nutrient solution for fertilization
10 - 300