Analytical Method: Analytical Quality Assurance, Standard for Total Chlorine
Reagents:
Cat.No. 102426  Chloramine T for analysis
Cat.No. 116754  Water for analysis
Preparation of a standard solution:
Dissolve 4.00 g of chloramine T with water for analysis in a calibrated or conformity-checked 1000-ml volumetric flask and make up to the mark with water for analysis. The stock solution prepared according to this procedure has a concentration of 1000 mg/l total chlorine.
Further investigational concentrations may be prepared from this stock solution by diluting accordingly.
Stability:
When stored in a cool place (refrigerator), the stock solution and the diluted investigational solutions remain stable for one day.
For use with test kits:
Measuring range
in mg/l Cl2
111174
MColortestTM Chlorine and pH Test (free + total chlorine)
0.1 - 1.5
111100
MColortestTM Chlorine and pH Test (free + total chlorine)
refill pack for 111174
111113
111114
111115
MColortestTM Chlorine Test Reagent Cl2-1 
MColortestTM Chlorine Test Reagent Cl2-2
MColortestTM Chlorine Test Reagent Cl2-3
114801
MColortestTM Chlorine Test (free + total chlorine, liquid)
0.1 - 2
114803
MColortestTM Chlorine Test (free + total chlorine, liquid)
refill pack for 114801
114826
MColortestTM Chlorine Test (free + total chlorine)
0.25 - 15
118326
MColortestTM Chlorine Test (free + total chlorine)
refill pack for 114826
For use with test kits:
Measuring range
in mg/l
116896
Reflectoquant® Chlorine Test (free + total chlorine)
0.5 - 10.0
100597
Spectroquant® Chlorine Cell Test (free + total chlorine)
0.03 - 6.00
100599
Spectroquant® Chlorine Test (free + total chlorine)
0.010 - 6.00
100602
Spectroquant® Chlorine Test (total chlorine)
0.010 - 6.00
100086
100087
100088
Spectroquant® Chlorine Reagents Cl2-1
Spectroquant® Chlorine Reagents Cl2-2
Spectroquant® Chlorine Reagents Cl2-3
0.010 - 6.00
119257
Spectroquant® Chlorine Powder Packs for non-Merck photometers for 10-ml-samples (total chlorine)
0 - 2.00
119258
Spectroquant® Chlorine Powder Packs for non-Merck photometers for 25-ml-samples (total chlorine)
0 - 10.00