Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Analytical Method: Calcium in Wine
Instruments and Reagents:
Cat.No. 173016  Spectroquant® VIS Spectrophotometer Prove 100 or
Cat.No. 173017  Spectroquant® UV/VIS Spectrophotometer Prove 300 or
Cat.No. 173018  Spectroquant® UV/VIS Spectrophotometer Prove 600 or
Cat.No. 109748  Spectroquant® Photometer NOVA 30 or
Cat.No. 109751  Spectroquant® Photometer NOVA 60
Cat.No. 114946  Rectangular cells 10 mm and/or
Cat.No. 114947  Rectangular cells 20 mm and/or
Cat.No. 114944  Rectangular cells 50 mm
Cat.No. 100858  Spectroquant® Calcium Cell Test
Cat.No. 108418  Titriplex® III for analysis
Cat.No. 116754  Water for analysis
Titriplex® III solution:
Dissolve 0.2 g Titriplex® III in 100 ml of water for analysis.
Sample preparation:
Analysis of white and rose wines: dilute the wine 1:100 with water for analysis.
Analysis of red wines: concerning the sample the procedure is the same as that for white wine. The difference is the blank solution: add 1 drop of Titriplex® III solution.
Analysis:
Determine with the above mentioned test kit.
Calculation:
Calcium content in mg/l Ca = analysis value in mg/l Ca x 100
Materials
Related Links
Categories