Analytical Method: COD in landfill seepage water
Instruments and Reagents:
Cat.No. 173016  Spectroquant® VIS Spectrophotometer Prove 100 or
Cat.No. 173017  Spectroquant® UV/VIS Spectrophotometer Prove 300 or
Cat.No. 173018  Spectroquant® UV/VIS Spectrophotometer Prove 600 or
Cat.No. 109748  Spectroquant® Photometer NOVA 30 or
Cat.No. 109751  Spectroquant® Photometer NOVA 60 or
Cat.No. 173632  Spectroquant® Colorimeter Move 100
Cat.No. 114946  Rectangular cells 10 mm and/or
Cat.No. 114947  Rectangular cells 20 mm and/or
Cat.No. 114944  Rectangular cells 50 mm
Cat.No. 114540  Spectroquant® COD Cell Test, measuring range 10 - 150 mg/l
Cat.No. 114895  Spectroquant® COD Cell Test, measuring range 15 - 300 mg/l
Cat.No. 114690  Spectroquant® COD Cell Test, measuring range 50 - 500 mg/l
Cat.No. 114541  Spectroquant® COD Cell Test, measuring range 25 - 1500 mg/l
Cat.No. 114691  Spectroquant® COD Cell Test, measuring range 300 - 3500 mg/l
Cat.No. 114555  Spectroquant® COD Cell Test, measuring range 500 - 10 000 mg/l
Cat.No. 101797  Spectroquant® COD Cell Test, measuring range 5000 - 90 000 mg/l
Cat.No. 116754  Water for analysis
Sample preparation:
Adaptation to the measuring range appropriate to the sample can be achieved by selecting the most suitable test. Higher concentrations of COD (> 90 000 mg/l) require dilution of the sample with COD-free water.
Analysis:
Determine with the above mentioned test kits.
Calculation:
COD content in mg/l COD = analysis value in mg/l COD
If the sample has been diluted with water, the result should be multiplied by the dilution factor.
Materials
Related Links
Categories