Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Analytical Method: Iron in soils
Instruments and Reagents:
Cat.No. 173016  Spectroquant® VIS Spectrophotometer Prove 100 or
Cat.No. 173017  Spectroquant® UV/VIS Spectrophotometer Prove 300 or
Cat.No. 173018  Spectroquant® UV/VIS Spectrophotometer Prove 600 or
Cat.No. 109748  Spectroquant® Photometer NOVA 30 or
Cat.No. 109751  Spectroquant® Photometer NOVA 60 or
Cat.No. 173632  Spectroquant® Colorimeter Move 100
Cat.No. 114946  Rectangular cells 10 mm and/or
Cat.No. 114947  Rectangular cells 20 mm and/or
Cat.No. 114944  Rectangular cells 50 mm
Cat.No. 114549  Spectroquant® Iron Cell Test
Cat.No. 114896  Spectroquant® Iron Cell Test*
Cat.No. 114761  Spectroquant® Iron Test
Cat.No. 100796  Spectroquant® Iron Test
Cat.No. 109970  Hydrochloric acid 1 mol/l for analysis
Cat.No. 100316  Hydrochloric acid 25 % for analysis
Cat.No. 105588  Sodium hydroxide solution 10 % for analysis
Cat.No. 109531  MColorpHastTM pH-indicator strips pH 0 - 6
Cat.No. 116754  Water for analysis
Charcoal activated for soil test
Folded filter
*no compatible with Move 100
Sample preparation:
In a glass beaker place 10 g of air-dried and sieved (mesh 2 mm) sample, with 100 ml hydrochloric acid
1 mol/l and a spatula-tipfull of charcoal activated for soil tests and mix well for 1 hour. The pH value should be
1 - 2 (check with pH-indicator strips). If necessary, adjust with hydrochloric acid 25 %.
Afterwards adjust the pH value of the suspension to 3.0 with sodium hydroxide solution 10 %, using a pH meter. Then remove the undissolved matter by filtration through a folded filter and wash the filter residues 3 times with water for analysis. Collect the filtrate and the washings in a 200-ml volumetric flask, finally fill up to volume with water for analysis and mix well.
Analysis:
Determine with the above mentioned test kits.
Calculation:
Iron content in mg/kg Fe = analysis value in mg/l Fe x 20