Analytical Method: Iron (total) in building materials
Instruments and Reagents:
Cat.No. 173016  Spectroquant® VIS Spectrophotometer Prove 100 or
Cat.No. 173017  Spectroquant® UV/VIS Spectrophotometer Prove 300 or
Cat.No. 173018  Spectroquant® UV/VIS Spectrophotometer Prove 600 or
Cat.No. 109748  Spectroquant® Photometer NOVA 30 or
Cat.No. 109751  Spectroquant® Photometer NOVA 60 or
Cat.No. 173632  Spectroquant® Colorimeter Move 100
Cat.No. 114946  Rectangular cells 10 mm and/or
Cat.No. 114947  Rectangular cells 20 mm and/or
Cat.No. 114944  Rectangular cells 50 mm
Cat.No. 114549  Spectroquant® Iron Cell Test
Cat.No. 114896  Spectroquant® Iron Cell Test*
Cat.No. 114761  Spectroquant® Iron Test
Cat.No. 100796  Spectroquant® Iron Test
Cat.No. 104885  Potassium hydrogen sulfate for analysis
Cat.No. 109535  MColorpHastTM Universal indicator strips pH 0 - 14
Cat.No. 116754  Water for analysis
*not compatible with Move 100
Sample preparation:
Place 200 mg of the finely powdered material along with 5 g potassium hydrogen sulfate in a DURAN test tube, mix and heat gently in a flame until a clear orange-colored melt is obtained and no more gas is given off. Allow to cool, add water for analysis to dissolve most of the residue and transfer the suspension to a 100-ml volumetric flask. Make up to the mark with water for analysis and mix well. Filter through a paper filter and adjust the pH to 1 - 7.
Analysis:
Determine with the above mentioned test kits.
Calculation:
Iron (total) content in % = analysis value in mg/l Fe x 0.05.