Analytical Method: Nickel in plating baths
Instruments and Reagents:
Cat.No. 173016  Spectroquant® VIS Spectrophotometer Prove 100 or
Cat.No. 173017  Spectroquant® UV/VIS Spectrophotometer Prove 300 or
Cat.No. 173018  Spectroquant® UV/VIS Spectrophotometer Prove 600 or
Cat.No. 109748  Spectroquant® Photometer NOVA 30 or
Cat.No. 109751  Spectroquant® Photometer NOVA 60 or
Cat.No. 173632  Spectroquant® Colorimeter Move 100
Cat.No. 114946  Rectangular cells 10 mm and/or
Cat.No. 114947  Rectangular cells 20 mm and/or
Cat.No. 114944  Rectangular cells 50 mm
Cat.No. 114554  Spectroquant® Nickel Cell Test
Cat.No. 114785  Spectroquant® Nickel Test
Cat.No. 116754  Water for analysis
Sample preparation:
In a 500-ml standard volumetric flask place 2.5 ml of bath solution, which has been cooled to room temperature, fill up to volume with water for analysis and mix well. Take from this solution a further 5 ml and place in a second 500-ml standard volumetric flask and again fill up with water for analysis and mix.
Determine readily the nickel content in solutions diluted in this fashion.
Analysis:
Determine with the above mentioned test kits
Calculation:
Nickel content in g/l = analysis value in mg/l Ni x 20
Materials
Related Links
Categories