Analytical Method: Phosphorus (total) in beer
Instruments and Reagents:
Cat.No. 173016  Spectroquant® VIS Spectrophotometer Prove 100 or
Cat.No. 173017  Spectroquant® UV/VIS Spectrophotometer Prove 300 or
Cat.No. 173018  Spectroquant® UV/VIS Spectrophotometer Prove 600 or
Cat.No. 109748  Spectroquant® Photometer NOVA 30 or
Cat.No. 109751  Spectroquant® Photometer NOVA 60 or
Cat.No. 173632  Spectroquant® Colorimeter Move 100
Cat.No. 114946  Rectangular cells 10 mm and/or
Cat.No. 114947  Rectangular cells 20 mm and/or
Cat.No. 114944  Rectangular cells 50 mm
Cat.No. 114543  Spectroquant® Phosphate Cell Test
Cat.No. 114729  Spectroquant® Phosphate Cell Test
Cat.No. 114848  Spectroquant® Phosphate Test
Cat.No. 100731  Sulfuric acid 95 - 97 % for analysis
Cat.No. 107209  Hydrogen peroxide 30 % (Perhydrol®) for analysis
Cat.No. 105590  Sodium hydroxide solution 32 %
Cat.No. 116754  Water for analysis
Sample preparation:
Degas 100 ml beer in an ultrasonic bath. Mix 50 ml of the carbon dioxide-free sample with 5 ml sulfuric acid
95 - 97 % in a 600-ml beaker and slowly evaporate on an electric plate until the solution becomes black. Allow to cool and then add Perhydrol® in 1-ml portions. Vigorous reaction accompanied with foaming will take place. Continue heating the mixture until the fume point is reached. If the solution becomes brown or black, add Perhydrol® until it again becomes colorless. In this example, approximately 30 ml Perhydrol® would be required for oxidation of the sample. Allow to cool, dilute with ca. 50 ml water for analysis in a volumetric flask, adjust to pH 5 - 7 with 14 - 15 ml sodium hydroxide solution 32 % using a pH-meter and make up to 1 l with water for analysis. Dilute this solution again 1+9 with water for analysis.
Analysis:
Determine with the above mentioned test kits.
Calculation:
Phosphorus (total) content in mg/l P = analysis result in mg/l P x 200
Procedural comparison:
The sample was analysed according to procedure § 35 LMBG (German Foodstuffs Law), section 31-00-6. 248 mg/l was determined. The above described procedure resulted in a concentration of 247 mg/l.