Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Analytical Method: Sulfite in shrimps
Instruments and Reagents:
Cat.No. 173016  Spectroquant® VIS Spectrophotometer Prove 100 or
Cat.No. 173017  Spectroquant® UV/VIS Spectrophotometer Prove 300 or
Cat.No. 173018  Spectroquant® UV/VIS Spectrophotometer Prove 600 or
Cat.No. 109748  Spectroquant® Photometer NOVA 30 or
Cat.No. 109751  Spectroquant® Photometer NOVA 60 or
Cat.No. 173632  Spectroquant® Colorimeter Move 100
Cat.No. 114946  Rectangular cells 10 mm and/or
Cat.No. 114947  Rectangular cells 20 mm and/or
Cat.No. 114944  Rectangular cells 50 mm
Cat.No. 101746  Spectroquant® Sulfite Test
Cat.No. 109535  MColorpHastTM Universal indicator strips pH 0 - 14
Cat.No. 137085  Sodium hydroxide solution 0.05 mol/l 
Cat.No. 108418  Titriplex® III
Cat.No. 116754  Water for analysis
Reagent 1:
Place about 50 ml 0.05 molar sodium hydroxide solution into a 100-ml Erlenmeyer flask. Subsequently dissolve 0.005 g Titriplex® III in it and make up to the mark with sodium hydroxide solution. Cool this solution with an EDTA concentration of 50 mg/l in the refrigerator or on ice to about 0 - 4°C.
Reaction:
In neutral solution, sulfite ions react with 2.2'-dinitro-5.5'-dithiodibenzoic acid (Ellman's reagent) to form an organic thiosulfate. This reaction results in the release of a thiol that is determined photometrically.
Measurement range:
1.0 - 60.0 mg/l SO32-
Sample preparation:
Homogenize 10 g of ready-to-cook shrimps and 50 ml of cooled sodium hydroxide/EDTA solution in a mixer for about 1 minute. Subsequently transfer the entire sample, with the aid of another 50 ml of sodium hydroxide/EDTA solution and 100 ml of ice-cooled water for analysis, to a 250-ml volumetric flask and stir on a magnetic stirrer for about 20 min. Any foam present should disappear. Subsequently make up to the mark with water for analysis and swirl the flask to homogenize. Then filter through a fluted filter, discarding the first few milliliters.
Note: The shrimps must be processed rapidly, as sulfite is quickly oxidized in air. Opened packages have only about ¼ of the original content after 24 hours.
Analysis:
The measurement is carried out with the above mentioned test set.
Calculation:
Measured value [mg/l] x 25 = sulfite content [mg/kg]       (kg and l equalized)
Results: Different shrimp samples were tested.
Sample Measured value
[mg/l] 
Spectroquant®
Recovery rate [%]
Shrimp 1 
40.2  -
Shrimp 1 spiked with 2 mg/l sulfite 
42.7 101.2
Shrimp 1 spiked with 4 mg/l sulfite 
44.3 100.2
Shrimp 1 spiked with 6 mg/l sulfite 
46.1 99.8
Shrimp 1 spiked with 8 mg/l sulfite 
48.0 99.6
Shrimp 1 spiked with 10 mg/l sulfite 
49.6 98.8
Sample Measured value
[mg/l] 
Spectroquant®
Recovery rate [%]
Shrimp 2 
37.8  -
Shrimp 2 spiked with 2 mg/l sulfite 
39.6 99.5
Shrimp 2 spiked with 4 mg/l sulfite 
40.3 96.4
Shrimp 2 spiked with 6 mg/l sulfite 
41.2 94.1
Shrimp 2 spiked with 8 mg/l sulfite 
45.2 98.7
Shrimp 2 spiked with 10 mg/l sulfite 
48.6 101.7
Sample Measured value
[mg/l] 
Spectroquant®
Comparative measurement
value [mg/l]
MColortest® Cat. 111148
Shrimp 3 measurement 1 
41.8 41.3
Shrimp 3 measurement 2 
42.6 40.6
Shrimp 3 measurement 3 
41.7 41.3