Analytical Method: Titanium in wastewater
Reagents:
Cat.No. 103053  N,N-Dimethylformamide
Cat.No. 106268  Sodium acetate for analysis
Cat.No. 100700  Thioglycolic acid for analysis
Cat.No. 116754  Water for analysis
2.3-Dihydroxynaphthaline, Aldrich Cat.-No. D11, 600-9
Membrane filter 0.45 μm pore diameter
Reagent 1:
Dissolve 2.5 g 2.3-dihydroxynaphthaline in 50 ml of dimethylformamide.
Reagent 2:
Sodium acetate solution, saturated at 25 °C.
Reagent 3:
Mix 40 g thioglycolic acid and 60 ml of water for analysis.
Measurement range:
0.25 - 7.00 mg/l Ti at cell length 10 mm and 405 nm
Sample preparation:
The sample solution must be clear; if not, filter through a fluted filter or membrane filter with 0.45 μm pore diameter.
Analysis:
Mix 5.0 ml of sample solution, 0.25 ml reagent 1, 1.0 ml reagent 2 and 0.25 ml reagent 3 in a test tube. Allow to stand for 5 minutes and then measure against water at 405 nm.
Parameter list for programming the photometer SQ 118
01 Method Ti
02 Cat.No. -------
03 Measuring range 0.25 - 7.00
04 Units mg/l
05 Reaction time 5 0 min
06 Cell 10 mm
07 Wavelength 405 nm
08 Calibration Factor
09 Evaluation Linear
10 Factor 2.65
Calculation when measuring the absorbance value using the photometer SQ118 without programming the method in the memory of the instrument:
Measured absorbance
of samples in A
Result in mg/l
[A sample * 2.65]
1.000 2.65
Materials
Related Links
Categories