TLC Analysis Analgesics on HPTLC Silica Gel 60 RP-18 WF254s (analyte set 3)
TLC Analysis Analgesics on HPTLC Silica Gel 60 RP-18 WF254s (analyte set 3)
Conditions
sample 0.2 µL
detector in situ evaluation with TLC/HPTLC scanner (Camag) UV 248 nm
migration distance 5 cm
plate HPTLC pre-coated plate RP-18 wF 254s (1.13124.0001)
chamber normal chamber without chamber saturation
eluent [A] acetonitrile; [B] water; (40:60, A:B)
Description
Categories Analytical Chromatography, TLC Applications, General TLC Applications, peptides
Featured Industry Pharmaceutical (small molecule)
suitability application for TLC