TLC Analysis of 4-Hydroxybenzoic Acid and 4-Hydroxybenzoic Acid Esters on HPTLC Silica Gel 60 RP-18 F254s: Effect of Eluent Stabilizer on Resolution
TLC Analysis of 4-Hydroxybenzoic Acid and 4-Hydroxybenzoic Acid Esters on HPTLC Silica Gel 60 RP-18 F254s: Effect of Eluent Stabilizer on Resolution
Conditions
sample 0.2 µL
detector in situ evaluation with TLC/HPTLC scanner (Camag) UV 254 nm
migration distance 7 cm
eluent chloroform with stabilizer, 2-methyl-2-butene or ethanol as shown
chamber normal chamber without chamber saturation
plate HPTLC pre-coated plate RP-18 F 254s (1.13724.0001)
Description
Categories Analytical Chromatography, TLC Applications, carboxylic acids
Featured Industry Chemicals and Industrial Polymers
General description TLC Plate Information: 1.13724.0001
suitability application for TLC