TLC Analysis of Alkaloids on HPTLC CN F254s
TLC Analysis of Alkaloids on HPTLC CN F254s
Conditions
sample 0.3 µl
detector in situ evaluation with TLC/HPTLC scanner (Camag) UV 254 nm
test parameters migration distance: 7 cm
test parameters eluent: [A] methanol; [B] l-propanol; [C] water; [D] ammonia
solution 25% (30:20:50:1, A:B:C:D) containing 0.1 M ammonium bromide
test parameters chamber: normal chamber without chamber saturation
test parameters plate: HPTLC pre-coated plate CN F 254s (1.16464.0001)
Description
Categories Analytical Chromatography, TLC Applications, peptides
Featured Industry Life Science and Biopharma
General description TLC Plate Information: 1.16464.0001
suitability application for TLC