Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
TLC Analysis of Analgesics on HPTLC Pre-coated Plate RP-18 WF254s (analyte set 1)
TLC Analysis of Analgesics on HPTLC Pre-coated Plate RP-18 WF254s (analyte set 1)
Conditions
sample 0.2 µl
detector in situ evaluation with TLC/HPTLC scanner (Camag) UV 254 nmdipped in dichlorofluorescein (0.2% in ethanol)
Description
Featured Industry Pharmaceutical (small molecule)
General description TLC Plate Information: 1.13124.0001
suitability application for TLC