TLC Analysis of Aromatic Acids by Ion-Pair Chromatography on HPTLC Silica Gel 60 RP-18 F254s
TLC Analysis of Aromatic Acids by Ion-Pair Chromatography on HPTLC Silica Gel 60 RP-18 F254s
Conditions
sample 0.3 µL
detector in situ evaluation with TLC/HPTLC scanner (Camag) UV 254 nm
migration distance 7 cm
plate TLC pre-coated plate RP-18 F254s (1.15423.0001)
eluent [A] acetone; [B] water; (50:50, A:B) containing 0.1 M tetraethylammoniumbromide
chamber normal chamber without chamber saturation
Description
Categories Analytical Chromatography, TLC Applications, organic acids
Featured Industry Chemicals and Industrial Polymers
General description TLC Plate Information: 1.15423.0001
suitability application for TLC