TLC Analysis of Barbiturates on HPTLC Silica Gel 60 RP-18 WF254s
TLC Analysis of Barbiturates on HPTLC Silica Gel 60 RP-18 WF254s
Conditions
sample 0.3 µLdipped in dichlorofluorescein (0.2% in ethanol)
detector in situ evaluation with TLC/HPTLC scanner (Camag) UV 248 nmdipped
in dichlorofluorescein (0.2% in ethanol)
migration distance 7 cm
plate HPTLC pre-coated plate RP-18 wF 254s (1.13124.0001, 1.13124.0001)
eluent [A] methanol; [B] water; (60:40, A:B)
chamber normal chamber without chamber saturation
Description
Categories Analytical Chromatography, TLC Applications, anxiolytics
Featured Industry Pharmaceutical (small molecule)
General description TLC Plate Information: 1.13124.0001
suitability application for TLC