TLC Analysis of Benzoic Acid and Sorbic Acid on HPTLC CN F254s (analyte set 1)
TLC Analysis of Benzoic Acid and Sorbic Acid on HPTLC CN F254s (analyte set 1)
Conditions
sample 0.2 µL
detector dipped in dichlorofluorescein (0.2% in ethanol) (in situ evaluation with TLC/HPTLC
scanner (Camag) UV 265 nm)
chamber normal chamber without chamber saturation
migration distance 7 cm
plate HPTLC pre-coated plate CN F 254s (1.16464.0001)
eluent [A] ethanol; [B] water; (20:80, A:B) containing 0.1 M tetraethylammonium chloride
Description
Categories Analytical Chromatography, TLC Applications, peptides
Featured Industry Life Science and Biopharma
suitability application for TLC