TLC Analysis of Dipeptides on ProteoChrom HPTLC Cellulose
TLC Analysis of Dipeptides on ProteoChrom HPTLC Cellulose
Conditions
detector spray reagent: ninhydrin, heated to 110 °C for 5 min
migration distance 7 cm
plate HPTLC pre-coated plate cellulose, without indicator (1.05651.0001, 1.16092.0001)
eluent [A] ethanol; [B] water; (80:20, A:B) containing 0.1 M lithium chloride
chamber normal chamber without chamber saturation
Description
Categories Analytical Chromatography, TLC Applications, peptides
Featured Industry Life Science and Biopharma
General description TLC Plate Information: 1.05651.0001
suitability application for TLC