TLC Analysis of DNP Amino Acids on HPTLC CN F254s
TLC Analysis of DNP Amino Acids on HPTLC CN F254s
Conditions
sample 0.4 µl
detector in situ evaluation with TLC/HPTLC scanner (Camag) UV 254 nm
test parameters eluent: [A] acetone; [B] acetone; [C] water; (45:10:45, A:B:C) containing 0.1 M lithium chloride
test parameters migration distance: 8 cm
test parameters chamber: normal chamber without chamber saturation
test parameters plate: HPTLC pre-coated plate CN F 254s (1.16464.0001)
Description
Categories Analytical Chromatography, TLC Applications, amino acids
Featured Industry Life Science and Biopharma
General description TLC Plate Information: 1.16464.0001
suitability application for TLC