TLC Analysis of Expectorants on HPTLC Silica Gel 60 RP-18 WF254s
TLC Analysis of Expectorants on HPTLC Silica Gel 60 RP-18 WF254s
Conditions
sample 0.3 µl
detector in situ evaluation with TLC/HPTLC scanner (Camag) UV 254 nm
test parameters eluent: [A] acetonitrile; [B] water; (30:70, A:B) with addition of 0.1 M TEACl
test parameters migration distance: 7 cm
test parameters plate: HPTLC pre-coated plate RP-18 wF 254s (1.13124.0001)
test parameters chamber: normal chamber without chamber saturation
Description
Categories Analytical Chromatography, TLC Applications, General TLC Applications
Featured Industry Pharmaceutical (small molecule)
General description TLC Plate Information: 1.13124.0001
suitability application for TLC