TLC Analysis of Fatty Acids on HPTLC Silica Gel 60 RP-18 F254s
TLC Analysis of Fatty Acids on HPTLC Silica Gel 60 RP-18 F254s
Conditions
sample 0.2 µL
detector dipped in dichlorofluorescein (0.2% ethanol)
migration distance 7 cm
eluent [A] ethanol; [B] water; (75:20, A:B)
chamber normal chamber without chamber saturation
plate HPTLC pre-coated plate RP-18 F 254s (1.13724.0001)
Description
Categories Analytical Chromatography, TLC Applications, fatty acids
Featured Industry Food and Beverages
Life Science and Biopharma
General description TLC Plate Information: 1.13724.0001
suitability application for TLC