TLC Analysis of FMOC-Peptides on HPTLC Silica Gel 60 RP-18 WF254s (analyte set 2)
TLC Analysis of FMOC-Peptides on HPTLC Silica Gel 60 RP-18 WF254s (analyte set 2)
Conditions
sample 0.3 µL
detector
in situ evaluation with TLC/HPTLC scanner (Camag) UV 254 nm
plate HPTLC pre-coated plate RP-18 wF 254s with concentrating zone (1.13124.0001)
chamber normal chamber without chamber saturation
migration distance 7 cm
eluent [A] acetonitrile; [B] water; (50:50, A:B) containing 0.1 M lithium chloride
Description
Categories Analytical Chromatography, TLC Applications, peptides
Featured Industry Life Science and Biopharma
suitability application for TLC